accessibility menu, dialog, popup

UserName

H3CNCH3

CAS RN 717880-39-2

IUPAC Name: 9,9-dimethyl-10-phenyl-9,10-dihydroacridine
Synonyms: 9,9-dimethyl-10-phenylacridine
Molecular Weight (g/mol): 285.39
Molecular Formula: C21H19N
InChi Key: CKTUEZIFXPDRMO-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2N(C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results