accessibility menu, dialog, popup

UserName

HBrCH3SNH2NH3C

CAS RN 7170-76-5

IUPAC Name: hydrogen dimethyl-1,3-thiazol-2-amine bromide
Synonyms: hydrogen dimethyl-1,3-thiazol-2-amine bromide
Molecular Weight (g/mol): 209.11
Molecular Formula: C5H9BrN2S
InChi Key: CATCJRCKBGAMKK-UHFFFAOYSA-N
SMILES: [H+].[Br-].CC1=C(C)N=C(N)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results