accessibility menu, dialog, popup

UserName

CAS RN 716-53-0

IUPAC Name: 9-chloroanthracene
Synonyms: 9-chloroanthracene
Molecular Weight (g/mol): 212.68
Molecular Formula: C14H9Cl
InChi Key: KULLJOPUZUWTMF-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results