accessibility menu, dialog, popup

UserName

CAS RN 7158-32-9

IUPAC Name: 4-[4-(carbonochloridoyl)phenoxy]benzoyl chloride
Synonyms: 4-[4-(carbonochloridoyl)phenoxy]benzoyl chloride
Molecular Weight (g/mol): 295.12
Molecular Formula: C14H8Cl2O3
InChi Key: OSUWBBMPVXVSOA-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(OC2=CC=C(C=C2)C(Cl)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results