accessibility menu, dialog, popup

UserName

CH3OOOHN

CAS RN 7149-65-7

IUPAC Name: ethyl 5-oxopyrrolidine-2-carboxylate
Synonyms: ethyl pca
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: QYJOOVQLTTVTJY-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCC(=O)N1
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)