accessibility menu, dialog, popup

UserName

H3CSOONOHOH

CAS RN 7146-67-0

IUPAC Name: N,N-bis(2-hydroxyethyl)-4-methylbenzene-1-sulfonamide
Synonyms: N,N-bis(2-hydroxyethyl)-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 259.32
Molecular Formula: C11H17NO4S
InChi Key: NHFJDXPINIIUQG-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N(CCO)CCO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results