accessibility menu, dialog, popup

UserName

HClH3CNH2ONHCH3H3C

CAS RN 71395-14-7

IUPAC Name: 2-amino-N-(2,6-dimethylphenyl)propanamide hydrochloride
Synonyms: tocainide hydrochloride
Molecular Weight (g/mol): 228.72
Molecular Formula: C11H17ClN2O
InChi Key: AMZACPWEJDQXGW-UHFFFAOYNA-N
SMILES: Cl.CC(N)C(=O)NC1=C(C)C=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results