accessibility menu, dialog, popup

UserName

ONOO

CAS RN 71311-82-5

IUPAC Name: 2-(4-nitrophenoxy)naphthalene
Synonyms: 2-(4-nitrophenoxy)naphthalene
Molecular Weight (g/mol): 265.27
Molecular Formula: C16H11NO3
InChi Key: ORYXOTYJSALBQZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC2=CC3=CC=CC=C3C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results