accessibility menu, dialog, popup

UserName

H3CSiH3CCH3OSiCH2OSiH3CCH3H3COSiCH3H3CCH3

CAS RN 7087-21-0

IUPAC Name: 2,2,6,6-tetramethyl-4-(prop-2-en-1-yl)-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptane
Synonyms: 2,2,6,6-tetramethyl-4-(prop-2-en-1-yl)-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptane
Molecular Weight (g/mol): 336.73
Molecular Formula: C12H32O3Si4
InChi Key: WCAXVXQTTCIZHD-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](CC=C)(O[Si](C)(C)C)O[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results