CAS RN 705-24-8
IUPAC Name:
4,6-dichloro-2-(trifluoromethyl)pyrimidine
Synonyms:
4,6-dichloro-2-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol):
216.97
Molecular Formula:
C5HCl2F3N2
InChi Key:
QFWVAJQVYBRTCL-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=NC(Cl)=CC(Cl)=N1
1
–
1
of
1
results
1
Promotions Available
4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%
4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%
CAS: 705-24-8 Molecular Formula: C5HCl2F3N2 Molecular Weight (g/mol): 216.97 MDL Number: MFCD08436597 InChI Key: QFWVAJQVYBRTCL-UHFFFAOYSA-N Synonym: 4,6-dichloro-2-trifluoromethyl pyrimidine,4,6-dichloro-2-trifluoromethyl-pyrimidine,pyrimidine, 4,6-dichloro-2-trifluoromethyl,pubchem19441,ksc496e6h,4 pound not6-dichloro-2-trifluoromethylpyrimidine PubChem CID: 15713197 IUPAC Name: 4,6-dichloro-2-(trifluoromethyl)pyrimidine SMILES: FC(F)(F)C1=NC(Cl)=CC(Cl)=N1