accessibility menu, dialog, popup

UserName

CAS RN 70484-02-5

BrNN

CAS RN 70484-02-5

IUPAC Name: 6-bromonaphthalene-2,3-dicarbonitrile
Synonyms: 6-bromonaphthalene-2,3-dicarbonitrile
Molecular Weight (g/mol): 257.09
Molecular Formula: C12H5BrN2
InChi Key: UERLNOIJNRUJKQ-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=C(C#N)C(=CC2=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results