accessibility menu, dialog, popup

UserName

CAS RN 704-15-4

NH2ONHOO

CAS RN 704-15-4

IUPAC Name: 1-(2-aminoacetyl)pyrrolidine-2-carboxylic acid
Synonyms: gly-pro
Molecular Weight (g/mol): 172.18
Molecular Formula: C7H12N2O3
InChi Key: KZNQNBZMBZJQJO-UHFFFAOYNA-N
SMILES: NCC(=O)N1CCCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results