missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-proline 98.0+%, TCI America™

Catalog No. G01371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01371G Supplier TCI America Supplier No. G01371G
Only null left

Specifications

Purity ≥98.0% (HPLC)
CAS 704-15-4
Color White
Content And Storage Refrigerator
Format Crystal-Powder at 20°C
Molecular Weight (g/mol) 172.18
Product Type Glycyl-L-proline
Quantity 1 g