accessibility menu, dialog, popup

UserName

CAS RN 7000-27-3

H3CO(R)O(R)OH(S)OH(S)HO(R)HO

CAS RN 7000-27-3

IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Synonyms: O1-methyl-glucose β-methylglucoside β-d-methylglucopyranoside M0779_sigma MGL methyl hexopyranoside β-methyl-d-glucoside methyl β-d-glucoside
Molecular Weight (g/mol): 194.18
Molecular Formula: C7H14O6
InChi Key: HOVAGTYPODGVJG-XUUWZHRGSA-N
SMILES: CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results