accessibility menu, dialog, popup

UserName

CAS RN 700-17-4

IUPAC Name: 2,3,5,6-tetrafluoroaniline
Synonyms: 2,3,5,6-tetrafluoroaniline
Molecular Weight (g/mol): 165.09
Molecular Formula: C6H3F4N
InChi Key: SPSWJTZNOXMMMV-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=CC(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results