accessibility menu, dialog, popup

UserName

CAS RN 69812-29-9

IUPAC Name: 2-acetamido-4-methyl-1,3-thiazole-5-sulfonyl chloride
Synonyms: 2-acetamido-4-methyl-1,3-thiazole-5-sulfonyl chloride
Molecular Weight (g/mol): 254.7
Molecular Formula: C6H7ClN2O3S2
InChi Key: NXGKPRKPUCSEIL-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC(C)=C(S1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results