CAS RN 69807-81-4
IUPAC Name:
1-(1-methyl-1H-pyrrol-2-yl)methanamine
Synonyms:
1-(1-methylpyrrol-2-yl)methanamine
Molecular Weight (g/mol):
110.16
Molecular Formula:
C6H10N2
InChi Key:
GGCBARJYVAPZJQ-UHFFFAOYSA-N
SMILES:
CN1C=CC=C1CN
1
–
2
of
2
results
1
(1-Methyl-1H-pyrrol-2-yl)methylamine, 97%, Thermo Scientific™
CAS: 69807-81-4 Molecular Formula: C6H10N2 Molecular Weight (g/mol): 110.16 MDL Number: MFCD02677708 InChI Key: GGCBARJYVAPZJQ-UHFFFAOYSA-N PubChem CID: 2776207 IUPAC Name: (1-methylpyrrol-2-yl)methanamine SMILES: CN1C=CC=C1CN