CAS RN 6975-60-6
IUPAC Name:
1-(furan-2-yl)propan-2-one
Synonyms:
2-propanone, 1-(2-furanyl)-
Molecular Weight (g/mol):
124.14
Molecular Formula:
C7H8O2
InChi Key:
IQOJTGSBENZIOL-UHFFFAOYSA-N
SMILES:
CC(=O)CC1=CC=CO1
1
–
1
of
1
results
1
Promotions Available
2-Furylacetone, 99%
2-Furylacetone, 99%
CAS: 6975-60-6 Molecular Formula: C7H8O2 Molecular Weight (g/mol): 124.139 MDL Number: MFCD02683084 InChI Key: IQOJTGSBENZIOL-UHFFFAOYSA-N Synonym: 2-furylacetone,furylacetone,2-acetonylfuran,2-propanone, 1-2-furanyl,furfuryl methyl ketone,1-2-furyl-2-propanone,methyl furfuryl ketone,1-2-furanyl-2-propanone,2-furfuryl methyl ketone,2-furyl-2-propanone PubChem CID: 228583 IUPAC Name: 1-(furan-2-yl)propan-2-one SMILES: CC(=O)CC1=CC=CO1