accessibility menu, dialog, popup

UserName

CH3OHH3C

CAS RN 6966-10-5

IUPAC Name: (3,4-dimethylphenyl)methanol
Synonyms: (3,4-dimethylphenyl)methanol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: OKGZCXPDJKKZAP-UHFFFAOYSA-N
SMILES: CC1=CC=C(CO)C=C1C
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results