accessibility menu, dialog, popup

UserName

CAS RN 696-60-6

IUPAC Name: 4-(aminomethyl)phenol
Synonyms: 4-hydroxybenzylamine
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: RQJDUEKERVZLLU-UHFFFAOYSA-N
SMILES: NCC1=CC=C(O)C=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)