accessibility menu, dialog, popup

UserName

BrO

CAS RN 6952-89-2

IUPAC Name: (4-bromophenyl)(cyclopropyl)methanone
Synonyms: (4-bromophenyl)(cyclopropyl)methanone
Molecular Weight (g/mol): 225.09
Molecular Formula: C10H9BrO
InChi Key: QTHHOINSCNBYQO-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C(=O)C1CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)