accessibility menu, dialog, popup

UserName

CAS RN 689-67-8

IUPAC Name: (5E)-6,10-dimethylundeca-5,9-dien-2-one
Synonyms: geranylacetone dihydropseudoionone
Molecular Weight (g/mol): 194.32
Molecular Formula: C13H22O
InChi Key: HNZUNIKWNYHEJJ-FMIVXFBMSA-N
SMILES: CC(C)=CCC\C(C)=C\CCC(C)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)