accessibility menu, dialog, popup

UserName

CH3OOCH3

CAS RN 68818-86-0

IUPAC Name: 9,10-diethoxyanthracene
Synonyms: 9,10-diethoxyanthracene
Molecular Weight (g/mol): 266.34
Molecular Formula: C18H18O2
InChi Key: GJNKQJAJXSUJBO-UHFFFAOYSA-N
SMILES: CCOC1=C2C=CC=CC2=C(OCC)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)