accessibility menu, dialog, popup

UserName

CAS RN 688-12-0

IUPAC Name: 2-(2-chloroacetamido)-4-methylpentanoic acid
Synonyms: 2-(2-chloroacetamido)-4-methylpentanoic acid
Molecular Weight (g/mol): 207.65
Molecular Formula: C8H14ClNO3
InChi Key: VDUNMYRYEYROFL-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)CCl)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results