accessibility menu, dialog, popup

UserName

CAS RN 68497-62-1

CH3H3CNHNONOH3CCH3OHSOHOO

CAS RN 68497-62-1

IUPAC Name: N-{2-[bis(propan-2-yl)amino]ethyl}-2-(2-oxopyrrolidin-1-yl)acetamide; sulfuric acid
Synonyms: pramiracetam
Molecular Weight (g/mol): 367.46
Molecular Formula: C14H29N3O6S
InChi Key: ACSROKXFXFNERX-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.CC(C)N(CCNC(=O)CN1CCCC1=O)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results