accessibility menu, dialog, popup

UserName

CAS RN 68401-88-7

H3CNCH3H3CH3CSSSSSSSSSSNi

CAS RN 68401-88-7

IUPAC Name: λ³-nickel(3+) bis((5-sulfanidyl-2-sulfanylidene-2H-1,3-dithiol-4-yl)sulfanide) tetrabutylazanium
Synonyms: λ³-nickel(3+) bis((5-sulfanidyl-2-sulfanylidene-1,3-dithiol-4-yl)sulfanide) tetrabutylammonium ion
Molecular Weight (g/mol): 693.83
Molecular Formula: C22H36NNiS10
InChi Key: RORVWUFVFQXASJ-UHFFFAOYSA-J
SMILES: [Ni+3].[S-]C1=C([S-])SC(=S)S1.[S-]C1=C([S-])SC(=S)S1.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results