accessibility menu, dialog, popup

UserName

CAS RN 683-98-7

IUPAC Name: methyl 2-bromo-2,2-difluoroacetate
Synonyms: methyl 2-bromo-2,2-difluoroacetate
Molecular Weight (g/mol): 188.96
Molecular Formula: C3H3BrF2O2
InChi Key: IOBCLXSGWMFVQJ-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results