accessibility menu, dialog, popup

UserName

CAS RN 6828-35-9

ClH2NI

CAS RN 6828-35-9

IUPAC Name: 5-chloro-2-iodoaniline
Synonyms: 5-chloro-2-iodoaniline
Molecular Weight (g/mol): 253.47
Molecular Formula: C6H5ClIN
InChi Key: FEOMAFDDLHSVMO-UHFFFAOYSA-N
SMILES: NC1=C(I)C=CC(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results