accessibility menu, dialog, popup

UserName

FClBr

CAS RN 68220-26-8

IUPAC Name: 2-(bromomethyl)-1-chloro-3-fluorobenzene
Synonyms: 2-(bromomethyl)-1-chloro-3-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: IGUVNNXFTDCASP-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Cl)=C1CBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results