accessibility menu, dialog, popup

UserName

CAS RN 68105-02-2

BrH3CNH3CCH3CH3

CAS RN 68105-02-2

IUPAC Name: dimethylditetradecylazanium bromide
Synonyms: dimethylditetradecylazanium bromide
Molecular Weight (g/mol): 518.75
Molecular Formula: C30H64BrN
InChi Key: IRMGVPILCPGYNQ-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results