missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldimyristylammonium Bromide 97.0+%, TCI America™

Catalog No. D235525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D235525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D235525G Supplier TCI America Supplier No. D235525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 68105-02-2
Melting Point 169°C
Color White
Molecular Formula C30H64BrN
Quantity 25 g
MDL Number MFCD00191429
Synonym Dimethylditetradecylammonium Bromide
InChI Key IRMGVPILCPGYNQ-UHFFFAOYSA-M
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCC
IUPAC Name dimethylditetradecylazanium bromide
Molecular Weight (g/mol) 518.75
PubChem CID 3017829
Formula Weight 518.75
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyldimyristylammonium Bromide
Show More Show Less
EINECSNumber (2)-0184&(9)-1971
TSCA No