accessibility menu, dialog, popup

UserName

H3C

CAS RN 67774-74-7

IUPAC Name: undecylbenzene
Synonyms: 1-phenylundecane
Molecular Weight (g/mol): 232.41
Molecular Formula: C17H28
InChi Key: XBEADGFTLHRJRB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results