accessibility menu, dialog, popup

UserName

H3COOClCl

CAS RN 6725-44-6

IUPAC Name: methyl 2-(3,4-dichlorophenyl)acetate
Synonyms: methyl 2-(3,4-dichlorophenyl)acetate
Molecular Weight (g/mol): 219.06
Molecular Formula: C9H8Cl2O2
InChi Key: FWVOAIHAIWHHDM-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results