accessibility menu, dialog, popup

UserName

CH3OSO

CAS RN 670-98-4

IUPAC Name: methyl benzenesulfinate
Synonyms: methyl benzenesulfinate
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: PSNSVDSRLUYDKF-UHFFFAOYNA-N
SMILES: COS(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)