accessibility menu, dialog, popup

UserName

CAS RN 67-16-3

IUPAC Name: N-[(4-amino-2-methylpyrimidin-5-yl)methyl]-N-[(2E)-3-{[(2E)-2-{N-[(4-amino-2-methylpyrimidin-5-yl)methyl]formamido}-5-hydroxypent-2-en-3-yl]disulfanyl}-5-hydroxypent-2-en-2-yl]formamide
Synonyms: N-[(4-amino-2-methylpyrimidin-5-yl)methyl]-N-[(2E)-3-{[(2E)-2-{N-[(4-amino-2-methylpyrimidin-5-yl)methyl]formamido}-5-hydroxypent-2-en-3-yl]disulfanyl}-5-hydroxypent-2-en-2-yl]formamide
Molecular Weight (g/mol): 562.71
Molecular Formula: C24H34N8O4S2
InChi Key: GFEGEDUIIYDMOX-YHARCJFQSA-N
SMILES: C\C(N(CC1=CN=C(C)N=C1N)C=O)=C(\CCO)SS\C(CCO)=C(/C)N(CC1=CN=C(C)N=C1N)C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results