CAS RN 669092-47-1
IUPAC Name:
4-[2-(4-aminophenyl)hydrazin-1-ylidene]-2-chlorocyclohexa-2,5-dien-1-one
Synonyms:
4-[2-(4-aminophenyl)hydrazin-1-ylidene]-2-chlorocyclohexa-2,5-dien-1-one
Molecular Weight (g/mol):
247.68
Molecular Formula:
C12H10ClN3O
InChi Key:
NWMGUTQRHABSPF-UHFFFAOYSA-N
SMILES:
NC1=CC=C(NN=C2C=CC(=O)C(Cl)=C2)C=C1
1
–
1
of
1
results
1
4-[(4-Aminophenyl)azo]-2-chlorophenol, 95%
CAS: 669092-47-1 Molecular Formula: C12H10ClN3O Molecular Weight (g/mol): 247.682 MDL Number: MFCD26401540 InChI Key: NWMGUTQRHABSPF-UHFFFAOYSA-N Synonym: e-4-4-aminophenyl diazenyl-2-chlorophenol,4-e-2-4-aminophenyl diazen-1-yl-2-chlorophenol PubChem CID: 86339119 IUPAC Name: 4-[(4-aminophenyl)hydrazinylidene]-2-chlorocyclohexa-2,5-dien-1-one SMILES: C1=CC(=CC=C1N)NN=C2C=CC(=O)C(=C2)Cl