CAS RN 666-52-4
CAS RN 666-52-4
IUPAC Name:
(²H₆)propan-2-one
Synonyms:
deuterated acetone
Molecular Weight (g/mol):
64.12
Molecular Formula:
C3H6O
InChi Key:
CSCPPACGZOOCGX-WFGJKAKNSA-N
SMILES:
[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]
1
–
11
of
11
results
1
Chloroform Solution NMR Reference Standard, 1 in acetone-d6 (99.9 atom D), NMR tube size 3 mm, MilliporeSigma™ Supelco™
Chloroform is a colorless liquid, which is volatile in nature. It is generally used as an inhalant anesthetic agent and also as an extraction solvent for oils, fats and greases.