CAS RN 6642-31-5
IUPAC Name:
6-amino-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
uracil, 6-amino-1,3-dimethyl-
Molecular Weight (g/mol):
155.16
Molecular Formula:
C6H9N3O2
InChi Key:
VFGRNTYELNYSKJ-UHFFFAOYSA-N
SMILES:
CN1C(N)=CC(=O)N(C)C1=O
1
–
1
of
1
results
1
6-Amino-1,3-dimethyluracil 98.0+%, TCI America™
CAS: 6642-31-5 Molecular Formula: C6H9N3O2 Molecular Weight (g/mol): 155.157 MDL Number: MFCD00006552 InChI Key: VFGRNTYELNYSKJ-UHFFFAOYSA-N PubChem CID: 81152 IUPAC Name: 6-amino-1,3-dimethylpyrimidine-2,4-dione SMILES: CN1C(=CC(=O)N(C1=O)C)N