accessibility menu, dialog, popup

UserName

H2NOFFFFFFF

CAS RN 662-50-0

IUPAC Name: heptafluorobutanamide
Synonyms: heptafluorobutanamide
Molecular Weight (g/mol): 213.06
Molecular Formula: C4H2F7NO
InChi Key: FOBJABJCODOMEO-UHFFFAOYSA-N
SMILES: NC(=O)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results