accessibility menu, dialog, popup

UserName

CAS RN 66185-74-8

IUPAC Name: 4-(hydroxymethyl)cyclohexane-1-carboxylic acid
Synonyms: 4-(hydroxymethyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: VQMIUUBKKPIDBN-UHFFFAOYSA-N
SMILES: OCC1CCC(CC1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results