accessibility menu, dialog, popup

UserName

NNBrBrBrBr

CAS RN 66127-00-2

IUPAC Name: 3,5,6,8-tetrabromo-1,10-phenanthroline
Synonyms: 3,5,6,8-tetrabromo-1,10-phenanthroline
Molecular Weight (g/mol): 495.79
Molecular Formula: C12H4Br4N2
InChi Key: BAPJVWLAFSQLNX-UHFFFAOYSA-N
SMILES: BrC1=CN=C2C(=C1)C(Br)=C(Br)C1=CC(Br)=CN=C21
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results