accessibility menu, dialog, popup

UserName

CAS RN 660-49-1

IUPAC Name: 3-fluoro-5-iodoaniline
Synonyms: 3-fluoro-5-iodoaniline
Molecular Weight (g/mol): 237.02
Molecular Formula: C6H5FIN
InChi Key: HTEPMXLKKDKDAQ-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results