accessibility menu, dialog, popup

UserName

CH3OClCl

CAS RN 6582-42-9

IUPAC Name: 1-(3,4-dichlorophenyl)propan-2-one
Synonyms: 1-(3,4-dichlorophenyl)propan-2-one
Molecular Weight (g/mol): 203.06
Molecular Formula: C9H8Cl2O
InChi Key: AJUAASWQUWIMHM-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(Cl)C(Cl)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)