accessibility menu, dialog, popup

UserName

CAS RN 654-53-5

FFFFFFF

CAS RN 654-53-5

IUPAC Name: 1,2,3,4-tetrafluoro-5-(trifluoromethyl)benzene
Synonyms: 1,2,3,4-tetrafluoro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 218.07
Molecular Formula: C7HF7
InChi Key: ZWVOHERGWLDGFT-UHFFFAOYSA-N
SMILES: FC1=CC(=C(F)C(F)=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results