accessibility menu, dialog, popup

UserName

CAS RN 65294-20-4

CH3H3CCH3H3CFFFFFF

CAS RN 65294-20-4

IUPAC Name: 4-[2-(3,4-dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-1,2-dimethylbenzene
Synonyms: 4-[2-(3,4-dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-1,2-dimethylbenzene
Molecular Weight (g/mol): 360.34
Molecular Formula: C19H18F6
InChi Key: GLFKFHJEFMLTOB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C)C(C1=CC=C(C)C(C)=C1)(C(F)(F)F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results