accessibility menu, dialog, popup

UserName

CAS RN 652154-13-7

N(R)PN(R)OPOOP(R)(S)OOOOOOOOHOHHONH2HONaNa

CAS RN 652154-13-7

IUPAC Name: disodium phosphono ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: disodium phosphono {[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol): 527.12
Molecular Formula: C9H14N3Na2O14P3
InChi Key: NFQMDTRPCFJJND-IJRYJGAHNA-L
SMILES: [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results