missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine 5'-triphosphate, disodium salt hydrate, 95%

Catalog No. AC226225000
Change view
Click to view available options
:
500mg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC226225000 500mg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC226225000 Supplier Thermo Scientific Chemicals Supplier No. 226225000
Only null left

CAS: 652154-13-7 | C9H18N3Na2O16P3 | 563.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Melting Point 215.0°C to 218.0°C
Color White
CAS 652154-13-7
Physical Form Crystalline Powder
Quantity 500 mg
Assay Percent Range 94% min. (HPLC)
Molecular Formula C9H18N3Na2O16P3
MDL Number MFCD00150807
Synonym cytidine 5'-triphosphate sodium salt, cytidine 5'-triphosphate tetrasodium
Solubility Information Solubility in water: 200g/L (20°C)
InChI Key KNSYTSUGTLJHPS-ODQFIEKDSA-L
SMILES O.O.[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O
IUPAC Name [[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
Molecular Weight (g/mol) 563.15
PubChem CID 129893565
Formula Weight 527.12
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Cytidine 5′-triphosphate,disodium salt hydrate, 95%
Show More Show Less

RUO – Research Use Only