accessibility menu, dialog, popup

UserName

H3COONOH

CAS RN 65214-82-6

IUPAC Name: ethyl 4-hydroxypiperidine-1-carboxylate
Synonyms: ethyl 4-hydroxypiperidine-1-carboxylate
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: QABJNOSERNVHDY-UHFFFAOYSA-N
SMILES: CCOC(=O)N1CCC(O)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results