accessibility menu, dialog, popup

UserName

H2NOFFFFF

CAS RN 652-31-3

IUPAC Name: pentafluorobenzamide
Synonyms: 2,3,4,5,6-pentafluorobenzamide
Molecular Weight (g/mol): 211.09
Molecular Formula: C7H2F5NO
InChi Key: WPWWHXPRJFDTTJ-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results