CAS RN 65100-04-1
IUPAC Name:
3-[diethoxy(methyl)silyl]propyl 2-methylprop-2-enoate
Synonyms:
3-[diethoxy(methyl)silyl]propyl 2-methylprop-2-enoate
Molecular Weight (g/mol):
260.41
Molecular Formula:
C12H24O4Si
InChi Key:
DOYKFSOCSXVQAN-UHFFFAOYSA-N
SMILES:
CCO[Si](C)(CCCOC(=O)C(C)=C)OCC
1
–
1
of
1
results
1
3-[Diethoxy(methyl)silyl]propyl Methacrylate 98.0+%, TCI America™
CAS: 65100-04-1 Molecular Formula: C12H24O4Si Molecular Weight (g/mol): 260.405 MDL Number: MFCD00053879 InChI Key: DOYKFSOCSXVQAN-UHFFFAOYSA-N Synonym: Methacrylic Acid 3-[Diethoxy(methyl)silyl]propyl Ester PubChem CID: 2759529 IUPAC Name: 3-[diethoxy(methyl)silyl]propyl 2-methylprop-2-enoate SMILES: CCO[Si](C)(CCCOC(=O)C(=C)C)OCC